COMH93D5ECB6 Combi-Blocks, Inc.

3-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-propyl-1,3-diazaspiro[4.4]non-1-en-4-one

  • MDL number MFCD20273012
  • Cas Number : 158778-58-6
  • Molecular Weight : 414.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8780

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8780
Availability details for
Quantity
  • Availability
  • Add to Cart