COMH93D5ECE6 Combi-Blocks, Inc.

N-(3-Bromophenyl)cyclopropanesulfonamide

  • MDL number MFCD20317864
  • Cas Number : 1235063-43-0
  • Molecular Weight : 276.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8837
Availability details for
Quantity
  • Availability
  • Add to Cart