COMH93D5ED06 Combi-Blocks, Inc.

4-Bromo-2-chloro-n-cyclopropylbenzamide

  • MDL number MFCD20358019
  • Cas Number : 893420-20-7
  • Molecular Weight : 274.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8874
Availability details for
Quantity
  • Availability
  • Add to Cart