COMH93D5ED18 Combi-Blocks, Inc.

Isobutyl (4-(hydroxymethyl)phenyl)carbamate

  • MDL number MFCD20414129
  • Cas Number : 1342997-75-4
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8897

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-8897
Availability details for
Quantity
  • Availability
  • Add to Cart