COMH93D5EDA0 Combi-Blocks, Inc.

N,N-Diethyl-2-formyl-6-methoxy-benzamide

  • MDL number MFCD20527268
  • Cas Number : 70946-17-7
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9066
Availability details for
Quantity
  • Availability
  • Add to Cart