COMH93D5EDCB Combi-Blocks, Inc.

2-Chloro-9h-purin-6-ol

  • MDL number MFCD20547732
  • Molecular Weight : 170.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9115
Availability details for
Quantity
  • Availability
  • Add to Cart