COMH93D5EE6A Combi-Blocks, Inc.

N-(3-Amino-4-hydroxyphenyl)acetamide

  • MDL number MFCD20691146
  • Cas Number : 102-33-0
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9312
Availability details for
Quantity
  • Availability
  • Add to Cart