COMH93D5EF18 Combi-Blocks, Inc.

tert-Butyl 2-methoxybenzylcarbamate

  • MDL number MFCD21105786
  • Cas Number : 153903-20-9
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9537
Availability details for
Quantity
  • Availability
  • Add to Cart