COMH93D5EF33 Combi-Blocks, Inc.

[(5-Bromo-2-methylphenyl)methyl](methyl)amine

  • MDL number MFCD21192999
  • Cas Number : 216873-88-0
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9569
Availability details for
Quantity
  • Availability
  • Add to Cart