COMH93D5EF34 Combi-Blocks, Inc.

[(5-Bromo-2-methylphenyl)methyl](ethyl)amine

  • MDL number MFCD21193000
  • Cas Number : 1520652-91-8
  • Molecular Weight : 228.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9570
Availability details for
Quantity
  • Availability
  • Add to Cart