COMH93D5EF49 Combi-Blocks, Inc.

[3-Bromo-4-(cyclobutylmethoxy)phenyl]methanol

  • MDL number MFCD21240244
  • Molecular Weight : 271.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9592

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9592
Availability details for
Quantity
  • Availability
  • Add to Cart