COMH93D5EF4C Combi-Blocks, Inc.

N-Ethyl-n-[(4-iodophenyl)methyl]cyclopropanamine

  • MDL number MFCD21243586
  • Molecular Weight : 301.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9595

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9595
Availability details for
Quantity
  • Availability
  • Add to Cart