COMH93D5EF5D Combi-Blocks, Inc.

[(3-Chloropyridin-4-yl)methyl](hexyl)amine

  • MDL number MFCD21286250
  • Cas Number : 1499899-64-7
  • Molecular Weight : 226.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9614
Availability details for
Quantity
  • Availability
  • Add to Cart