COMH93D5EF8E Combi-Blocks, Inc.

N-Alpha-(9-fluorenylmethyloxycarbonyl)-n-epsilon-[15-(biotinamido)-4,7,10,13-tetraoxa-pentadecanoyl]-l-lysine

  • MDL number MFCD21363320
  • Cas Number : 1334172-64-3
  • Molecular Weight : 842.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9675
Availability details for
Quantity
  • Availability
  • Add to Cart