COMH93D5EF9D Combi-Blocks, Inc.

2-Methoxy-5-(pyrrolidin-3-yl)pyridine hydrochloride

  • MDL number MFCD21364632
  • Cas Number : 1359656-04-4
  • Molecular Weight : 214.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9691
Availability details for
Quantity
  • Availability
  • Add to Cart