COMH93D5EFFC Combi-Blocks, Inc.

Ethyl 3-amino-2-(3,4-dimethoxyphenyl)propanoate hcl

  • MDL number MFCD21647717
  • Cas Number : 16226-24-7
  • Molecular Weight : 289.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9804
Availability details for
Quantity
  • Availability
  • Add to Cart