COMH93D5F059 Combi-Blocks, Inc.

5-Bromo-3-chloro-2-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD21977703
  • Cas Number : 1289112-68-0
  • Molecular Weight : 261.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-9911
Availability details for
Quantity
  • Availability
  • Add to Cart