COMH93D5F0CF Combi-Blocks, Inc.

tert-Butyl 1-(3-(benzyloxy)-2-hydroxypropyl)-1h-1,2,3-triazole-4-carboxylate

  • MDL number MFCD30472058
  • Cas Number : 1851932-54-1
  • Molecular Weight : 333.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0019
Availability details for
Quantity
  • Availability
  • Add to Cart