COMH93D5F0DE Combi-Blocks, Inc.

(4-Methylmorpholin-3-yl)methanol hydrochloride

  • MDL number MFCD29920589
  • Cas Number : 1956379-95-5
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0036
Availability details for
Quantity
  • Availability
  • Add to Cart