COMH93D5F0E9 Combi-Blocks, Inc.

D-Glutamic acid gamma-tertbutyl ester alpha-methyl ester

  • MDL number MFCD23161864
  • Cas Number : 121069-22-5
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0047
Availability details for
Quantity
  • Availability
  • Add to Cart