COMH93D5F0FB Combi-Blocks, Inc.

3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-[2-(sulfooxy)ethyl]-thiazolium chloride

  • MDL number MFCD27966927
  • Cas Number : 15743-04-1
  • Molecular Weight : 380.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0069
Availability details for
Quantity
  • Availability
  • Add to Cart