COMH93D5F108 Combi-Blocks, Inc.

(2,3,4-Trifluoro-5-hydroxyphenyl)boronic acid

  • MDL number MFCD31652452
  • Cas Number : 2304633-92-7
  • Molecular Weight : 191.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0086
Availability details for
Quantity
  • Availability
  • Add to Cart