COMH93D5F10C Combi-Blocks, Inc.

2-(3-Bromo-4-fluorophenyl)acetohydrazide

  • MDL number MFCD30209678
  • Cas Number : 1570562-61-6
  • Molecular Weight : 247.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0092
Availability details for
Quantity
  • Availability
  • Add to Cart