COMH93D5F10D Combi-Blocks, Inc.

1-(2-Fluoro-4-methylphenyl)pyrrolidin-2-one

  • MDL number MFCD31654402
  • Cas Number : 2279122-99-3
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0093
Availability details for
Quantity
  • Availability
  • Add to Cart