COMH93D5F117 Combi-Blocks, Inc.

H-Beta-hoglu(otbu)-oh

  • MDL number MFCD23381247
  • Cas Number : 1275612-13-9
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0107
Availability details for
Quantity
  • Availability
  • Add to Cart