COMH93D5F12B Combi-Blocks, Inc.

5-Chloro-2-ethyl-4h-chromen-4-one

  • MDL number MFCD30471945
  • Cas Number : 1899895-07-8
  • Molecular Weight : 208.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0132
Availability details for
Quantity
  • Availability
  • Add to Cart