COMH93D5F153 Combi-Blocks, Inc.

7-Bromo-3-hydroxyisoindolin-1-one

  • MDL number MFCD28359529
  • Cas Number : 1936016-08-8
  • Molecular Weight : 228.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0184

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0184
Availability details for
Quantity
  • Availability
  • Add to Cart