COMH93D5F157 Combi-Blocks, Inc.

1-Bromo-4-(2,2-difluoroethoxy)-2-methylbenzene

  • MDL number MFCD25995408
  • Cas Number : 1547928-53-9
  • Molecular Weight : 251.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0191

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0191
Availability details for
Quantity
  • Availability
  • Add to Cart