COMH93D5F15D Combi-Blocks, Inc.

4'-(Acetoxymethyl)-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD30529571
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0199
Availability details for
Quantity
  • Availability
  • Add to Cart