COMH93D5F15E Combi-Blocks, Inc.

3-N-Di-boc-2-methyl-propionic acid

  • MDL number MFCD24475052
  • Cas Number : 357610-29-8
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0200
Availability details for
Quantity
  • Availability
  • Add to Cart