COMH93D5F16D Combi-Blocks, Inc.

3-Amino-n-benzyl-5-methylbenzamide

  • MDL number MFCD27049399
  • Cas Number : 1878808-42-4
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0217
Availability details for
Quantity
  • Availability
  • Add to Cart