COMH93D5F185 Combi-Blocks, Inc.

2-(3,5-Dimethyl-1h-1,2,4-triazol-1-yl)ethanamine hydrochloride

  • MDL number MFCD28142411
  • Cas Number : 1993128-72-5
  • Molecular Weight : 176.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0246
Availability details for
Quantity
  • Availability
  • Add to Cart