COMH93D5F1BE Combi-Blocks, Inc.

4-Iodo-3-(trifluoromethyl)pyridine

  • MDL number MFCD26516644
  • Cas Number : 1412904-86-9
  • Molecular Weight : 273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0314
Availability details for
Quantity
  • Availability
  • Add to Cart