COMH93D5F1F9 Combi-Blocks, Inc.

3-(4-Methylpiperazin-1-yl)-5-nitrobenzoic acid

  • MDL number MFCD30179626
  • Cas Number : 851528-03-5
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0384
Availability details for
Quantity
  • Availability
  • Add to Cart