COMH93D5F240 Combi-Blocks, Inc.

Ethyl 3-hydroxy-1-(4-(trifluoromethyl)phenyl)-1h-pyrazole-4-carboxylate

  • MDL number MFCD29922336
  • Cas Number : 675148-19-3
  • Molecular Weight : 300.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0468
Availability details for
Quantity
  • Availability
  • Add to Cart