COMH93D5F256 Combi-Blocks, Inc.

7-Bromo-2-phenylthieno[3,2-c]pyridin-4(5h)-one

  • MDL number MFCD29044859
  • Cas Number : 690636-04-5
  • Molecular Weight : 306.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0495
Availability details for
Quantity
  • Availability
  • Add to Cart