COMH93D5F257 Combi-Blocks, Inc.

3-Bromoquinoline-8-carbonitrile

  • MDL number MFCD30151096
  • Cas Number : 1699370-11-0
  • Molecular Weight : 233.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0496
Availability details for
Quantity
  • Availability
  • Add to Cart