COMH93D5F265 Combi-Blocks, Inc.

Methyl 2-(chlorosulfonyl)-4-nitrobenzoate

  • MDL number MFCD27980731
  • Cas Number : 154712-47-7
  • Molecular Weight : 279.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0512

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0512
Availability details for
Quantity
  • Availability
  • Add to Cart