COMH93D5F367 Combi-Blocks, Inc.

9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene

  • MDL number MFCD30470615
  • Cas Number : 844679-02-3
  • Molecular Weight : 409.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0825
Availability details for
Quantity
  • Availability
  • Add to Cart