COMH93D5F378 Combi-Blocks, Inc.

4-Bromo-2-(difluoromethyl)-6-methylpyridine

  • MDL number MFCD25477799
  • Cas Number : 1226800-12-9
  • Molecular Weight : 222.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0848

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0848
Availability details for
Quantity
  • Availability
  • Add to Cart