COMH93D5F3A3 Combi-Blocks, Inc.

5-Chloro-4-methyl-2-propoxyaniline

  • MDL number MFCD28580320
  • Cas Number : 1369208-23-0
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0904
Availability details for
Quantity
  • Availability
  • Add to Cart