COMH93D5F3A6 Combi-Blocks, Inc.

N,N'-Dme-n,n'-bis-peg2-acid

  • MDL number MFCD30536147
  • Cas Number : 2062663-61-8
  • Molecular Weight : 408.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0907
Availability details for
Quantity
  • Availability
  • Add to Cart