COMH93D5F3CC Combi-Blocks, Inc.

2-(1,3-Dioxolan-2-yl)ethanol

  • MDL number MFCD25964441
  • Cas Number : 5465-08-7
  • Molecular Weight : 118.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-0956
Availability details for
Quantity
  • Availability
  • Add to Cart