COMH93D5F3FC Combi-Blocks, Inc.

Methyl 2-(4-bromo-3-[(cyclopropylamino)methyl]phenoxy)acetate

  • MDL number MFCD25634251
  • Cas Number : 1521001-50-2
  • Molecular Weight : 314.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1010
Availability details for
Quantity
  • Availability
  • Add to Cart