COMH93D5F3FE Combi-Blocks, Inc.

(E)-3-((1-Methyl-1h-indol-3-yl)methylene)-1h-pyrrolo[3,2-b]pyridin-2(3h)-one

  • MDL number MFCD22631496
  • Cas Number : 1243657-78-4
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1014
Availability details for
Quantity
  • Availability
  • Add to Cart