COMH93D5F41F Combi-Blocks, Inc.

1-(5-Bromo-2-fluoro-benzyl)-4-ethoxy-1h-pyrazole

  • MDL number MFCD31654523
  • Cas Number : 2270911-91-4
  • Molecular Weight : 299.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1056
Availability details for
Quantity
  • Availability
  • Add to Cart