COMH93D5F484 Combi-Blocks, Inc.

Methyl 7-(4-(dimethoxymethyl)phenylcarbamoyl)heptanoate

  • MDL number MFCD22381074
  • Cas Number : 910610-25-2
  • Molecular Weight : 337.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1180
Availability details for
Quantity
  • Availability
  • Add to Cart