COMH93D5F497 Combi-Blocks, Inc.

(R)-N-(2-Aminopropyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD31652653
  • Cas Number : 2208822-46-0
  • Molecular Weight : 170.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1208
Availability details for
Quantity
  • Availability
  • Add to Cart