COMH93D5F4DA Combi-Blocks, Inc.

1,3-Bis(trifluoromethyl)cyclohexane

  • MDL number MFCD31009310
  • Cas Number : 79850-12-7
  • Molecular Weight : 220.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1289
Availability details for
Quantity
  • Availability
  • Add to Cart