COMH93D5F4ED Combi-Blocks, Inc.

Capsaicin beta-d-glucopyranoside

  • MDL number MFCD22124716
  • Cas Number : 153409-16-6
  • Molecular Weight : 467.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-1312
Availability details for
Quantity
  • Availability
  • Add to Cart